2,2'-Bipyridin-4,4'-dicarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4,4'-Dicarboxy-2,2'-bipyridine, [2,2'-Bipyridine]-4,4'-dicarboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

550566-1G
SIAL550566-1GEA 42.8 EUR
SIAL550566-1G
2,2'-Bipyridin-4,4'-dicarbonsäure, Sigma-Aldrich®
2,2'-Bipyridin-4,4'-dicarbonsäure
Formel: C₁₂H₈N₂O₄
Molecular Weight: 244,20 g/mol
MDL: MFCD00015430
CAS-Nummer: 6813-38-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR