2-Nitro-4-thiocyanatoanilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-Amino-3-nitro-phenyl) thiocyanate

M-Clarity™ Qualitätsstufe = MQ100

550647-25G
SIAL550647-25GEA 0 EUR
SIAL550647-25G
2-Nitro-4-thiocyanatoanilin, Sigma-Aldrich®
2-Nitro-4-thiocyanatoanilin
Formel: C₇H₅N₃O₂S
Molecular Weight: 195,2000 g/mol
MDL: MFCD00463878
CAS-Nummer: 54029-45-7
EINECS: 258-931-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR