Sigma-Aldrich® One-piece fingertight fittings
Lieferant: Merck
55067-U
SIAL55067-UEA 120 EUR
SIAL55067-U
Sigma-Aldrich® One-piece fingertight fittings
Fittings und Verbinder
Standard knurled PEEK fittings with 10-32 thread, no ferrule to drop onto the floor.