3-Iodbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

550760-1G
SIAL550760-1GEA 43.9 EUR
SIAL550760-1G
3-Iodbenzaldehyd, Sigma-Aldrich®
3-Iodobenzaldehyde
Formel: C₇H₅IO
Molecular Weight: 232,02 g/mol
MDL: MFCD00039573
CAS-Nummer: 696-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR