2-Iodbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

550779-1G
SIAL550779-1GEA 147 EUR
SIAL550779-1G
2-Iodbenzaldehyd, Sigma-Aldrich®
2-Iodbenzaldehyd
Formel: C₇H₅IO
Molecular Weight: 232,02 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00039570
CAS-Nummer: 26260-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR