2,4-Dichlor-6,7-dimethoxychinazolin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
550868-25G
SIAL550868-25GEA
160
EUR
SIAL550868-25G
2,4-Dichlor-6,7-dimethoxychinazolin, Sigma-Aldrich®
2,4-Dichloro-6,7-dimethoxyquinazoline
Formel:
C₁₀H₈Cl₂N₂O₂ Molecular Weight: 259,0887 g/mol |
MDL:
MFCD00051733 CAS-Nummer: 27631-29-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...