8-Quinolinol hemisulphate salt, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 8-Hydroxyquinoline hemisulphate, 8-Quinolinone hemisulphate, Oxine sulphate, Quinolin-8-ol hemisulphate, 8-Quinolinol sulphate, 8-Hydroxyquinolinium hemisulphate, Chinosol

55100-500G-F
SIAL55100-500G-FEA 151 EUR
SIAL55100-500G-F
8-Quinolinol hemisulphate salt, Sigma-Aldrich®
8-Quinolinol hemisulphate salt
Formel: C₁₈H₁₄N₂O₂·H₂SO₄
Molecular Weight: 388,39 g/mol
Schmelzpunkt: 177,5 °C
MDL: MFCD00013095
CAS-Nummer: 134-31-6
EINECS: 205-137-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR