(3-(Methylamino)propyl)trimethoxysilan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Trimethoxy(3-(methylamino)propyl)silan, N-Methyl-3-(trimethoxysilyl)propylamin, (3-Methylaminopropyl)trimethoxysilan
M-Clarity™ Qualitätsstufe = MQ200
551635-25ML
SIAL551635-25MLEA
123
EUR
SIAL551635-25ML
(3-(Methylamino)propyl)trimethoxysilan, Sigma-Aldrich®
(3-(Methylamino)propyl)trimethoxysilan
Formel:
H₃CNHCH₂CH₂CH₂Si(OCH₃)₃ Molecular Weight: 193,32 g/mol |
MDL:
MFCD00048021 CAS-Nummer: 3069-25-8 EINECS: 221-334-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...