2-Hydroxy-3-nitro-5-picolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Methyl-3-nitro-2-pyridon, 2-Hydroxy-5-methyl-3-nitropyridin

M-Clarity™ Qualitätsstufe = MQ100

551910-5G
SIAL551910-5GEA 34 EUR
SIAL551910-5G
2-Hydroxy-3-nitro-5-picolin, Sigma-Aldrich®
2-Hydroxy-3-nitro-5-picolin
Formel: C₆H₆N₂O₃
Molecular Weight: 154,13 g/mol
MDL: MFCD02070021
CAS-Nummer: 7464-14-4
EINECS: 244-647-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR