3-(1,1,2,2-Tetrafluorethoxy)benzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

552208-5G
SIAL552208-5GEA 89 EUR
SIAL552208-5G
3-(1,1,2,2-Tetrafluorethoxy)benzoesäure, Sigma-Aldrich®
3-(1,1,2,2-Tetrafluoroethoxy)benzoic acid
Formel: C₉H₆F₄O₃
MDL: MFCD00155925
CAS-Nummer: 70126-48-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR