2-Fluor-5-iodbenzoesäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

552437-5G
SIAL552437-5GEA 80.6 EUR
SIAL552437-5G
2-Fluor-5-iodbenzoesäure, Sigma-Aldrich®
2-Fluor-5-iodbenzoesäure
Formel: C₇H₄FIO₂
Molecular Weight: 266,01 g/mol
MDL: MFCD03094517
CAS-Nummer: 124700-41-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR