2-Fluor-6-iodbenzoylchlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

552445-5G
SIAL552445-5GEA 314 EUR
SIAL552445-5G
2-Fluor-6-iodbenzoylchlorid, Sigma-Aldrich®
2-Fluor-6-iodbenzoylchlorid
Formel: C₇H₃ClFIO
Molecular Weight: 284,46 g/mol
MDL: MFCD03094410
CAS-Nummer: 111771-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR