2-Fluor-5-iodbenzoylchlorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
552461-5G
SIAL552461-5GEA
103
EUR
SIAL552461-5G
2-Fluor-5-iodbenzoylchlorid, Sigma-Aldrich®
2-Fluor-5-iodbenzoylchlorid
Formel:
C₇H₃ClFIO |
MDL:
MFCD03094145 CAS-Nummer: 186584-73-6 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...