5-Amino-2-brompyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2-Brompyridin-5-amin, 2-Brom-5-pyridylamin

M-Clarity™ Qualitätsstufe = MQ200

552844-5G
SIAL552844-5GEA 39.2 EUR
SIAL552844-5G
5-Amino-2-brompyridin, Sigma-Aldrich®
5-Amino-2-brompyridin
Formel: C₅H₅BrN₂
Molecular Weight: 173.01 g/mol
MDL: MFCD01646116
CAS-Nummer: 13534-97-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR