1-Butyl-3-methylimidazoliumtetrachloroaluminat, Sigma-Aldrich®

Lieferant: Merck

55292-100G-F
SIAL55292-100G-FEA 206 EUR
SIAL55292-100G-F
1-Butyl-3-methylimidazoliumtetrachloroaluminat, Sigma-Aldrich®
1-Butyl-3-methylimidazoliumtetrachloroaluminate
Formel: C₈H₁₅AlCl₄N₂
Molecular Weight: 308.01 g/mol
CAS-Nummer: 80432-09-3
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR