Hafniumtetrakis[ethyl(methyl)azanid], Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

553123-25ML 553123-5ML
SIAL553123-25MLEA 572 EUR
SIAL553123-25ML SIAL553123-5ML
Hafniumtetrakis[ethyl(methyl)azanid], Sigma-Aldrich®
Tetrakis(ethylmethylamino)hafnium(IV)
Formel: C₁₂H₃₂HfN₄
MDL: MFCD03427130
CAS-Nummer: 352535-01-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR