Tetrakis(ethylmethylamino)zirkonium(IV), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

553131-5G
SIAL553131-5GEA 361 EUR
SIAL553131-5G
Tetrakis(ethylmethylamino)zirkonium(IV), Sigma-Aldrich®
Tetrakis(ethylmethylamino)zirkonium(IV)
Formel: C₁₂H₃₂N₄Zr
MDL: MFCD03427131
CAS-Nummer: 175923-04-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR