1-Brom-4-(difluormethoxy)benzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Difluoromethoxy)bromobenzene

M-Clarity™ Qualitätsstufe = MQ100

553425-1G
SIAL553425-1GEA 26 EUR
SIAL553425-1G
1-Brom-4-(difluormethoxy)benzol, Sigma-Aldrich®
1-Brom-4-(difluormethoxy)benzol
Formel: C₇H₅BrF₂O
Molecular Weight: 223,02 g/mol
MDL: MFCD00221465
CAS-Nummer: 5905-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR