1-Brom-3-(difluormethoxy)benzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

553433-1G
SIAL553433-1GEA 30.1 EUR
SIAL553433-1G
1-Brom-3-(difluormethoxy)benzol, Sigma-Aldrich®
1-Brom-3-(difluormethoxy)benzol
Formel: C₇H₅BrF₂O
Molecular Weight: 223,02 g/mol
MDL: MFCD00236233
CAS-Nummer: 262587-05-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR