6-Brom-3-methoxysalicylaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Brom-2-hydroxy-3-methoxybenzaldehyd

553581-25G
SIAL553581-25GEA 395 EUR
SIAL553581-25G
6-Brom-3-methoxysalicylaldehyd, Sigma-Aldrich®
6-Brom-3-methoxysalicylaldehyd
Formel: C₈H₇BrO₃
Molecular Weight: 231,05 g/mol
MDL: MFCD00046152
CAS-Nummer: 20035-41-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR