4'-Methoxy-3'-nitroacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(4-Methoxy-3-nitrophenyl)ethanone

M-Clarity™ Qualitätsstufe = MQ100

553654-10G
SIAL553654-10GEA 74 EUR
SIAL553654-10G
4'-Methoxy-3'-nitroacetophenon, Sigma-Aldrich®
4'-Methoxy-3'-nitroacetophenon
Formel: C₉H₉NO₄
Molecular Weight: 195,18 g/mol
MDL: MFCD00017023
CAS-Nummer: 6277-38-9
EINECS: 228-476-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR