4'-Brom-3'-nitroacetophenon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-(4-Brom-3-nitrophenyl)ethan-1-on
M-Clarity™ Qualitätsstufe = MQ100
553662-5G
SIAL553662-5GEA
53.8
EUR
SIAL553662-5G
4'-Brom-3'-nitroacetophenon, Sigma-Aldrich®
4'-Brom-3'-nitroacetophenon
Formel:
C₈H₆BrNO₃ Molecular Weight: 244,04 g/mol Schmelzpunkt: 120 °C |
MDL:
MFCD00016985 CAS-Nummer: 18640-58-9 EINECS: 242-469-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...