1,1-Cyclohexandiessigsäuremonoamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(1-(2-Amino-2-oxoethyl)cyclohexyl)acetic acid, 2-[1-(2-Amino-2-oxoethyl)cyclohexyl]acetic Acid

M-Clarity™ Qualitätsstufe = MQ100

553735-25G
SIAL553735-25GEA 209 EUR
SIAL553735-25G
1,1-Cyclohexandiessigsäuremonoamid, Sigma-Aldrich®
1,1-Cyclohexandiessigsäuremonoamid
Formel: C₁₀H₁₇NO₃
Molecular Weight: 199,2500 g/mol
MDL: MFCD02181086
CAS-Nummer: 99189-60-3
EINECS: 449-430-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR