2,5-Difluorbenzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,5-Difluorbenzolsulfonsäureamid

M-Clarity™ Qualitätsstufe = MQ100

553875-1G
SIAL553875-1GEA 22.8 EUR
SIAL553875-1G
2,5-Difluorbenzolsulfonamid, Sigma-Aldrich®
2,5-Difluorbenzolsulfonamid
Formel: C₆H₅F₂NO₂S
Molecular Weight: 193,17 g/mol
MDL: MFCD00798115
CAS-Nummer: 120022-63-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR