2,4-Difluorbenzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4-Difluorbenzolsulfonsäureamid

M-Clarity™ Qualitätsstufe = MQ100

553883-1G
SIAL553883-1GEA 24.3 EUR
SIAL553883-1G
2,4-Difluorbenzolsulfonamid, Sigma-Aldrich®
2,4-Difluorbenzolsulfonamid
Formel: C₆H₅F₂NO₂S
Molecular Weight: 193,17 g/mol
MDL: MFCD01320743
CAS-Nummer: 13656-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR