(2,4-Dimethoxyphenyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2,4-Dimethoxyphenylacetic Acid

M-Clarity™ Qualitätsstufe = MQ200

554197-5G
SIAL554197-5GEA 68.5 EUR
SIAL554197-5G
(2,4-Dimethoxyphenyl)essigsäure, Sigma-Aldrich®
(2,4-Dimethoxyphenyl)essigsäure
Formel: (C₆H₃(OCH₃)₂)CH₂COOH
Molecular Weight: 196,2 g/mol
MDL: MFCD00183328
CAS-Nummer: 6496-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR