4-Fluorsalicylsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Fluoro-2-hydroxybenzoic acid

M-Clarity™ Qualitätsstufe = MQ100

554332-1G
SIAL554332-1GEA 30.9 EUR
SIAL554332-1G
4-Fluorsalicylsäure, Sigma-Aldrich®
4-Fluorsalicylsäure
Formel: C₇H₅FO₃
Molecular Weight: 156,11 g/mol
MDL: MFCD00153168
CAS-Nummer: 345-29-9
EINECS: 206-459-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR