(R)-(+)-4-Benzyl-5,5-diphenyl-2-oxazolidinon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

554502-1G
SIAL554502-1GEA 151 EUR
SIAL554502-1G
(R)-(+)-4-Benzyl-5,5-diphenyl-2-oxazolidinon, Sigma-Aldrich®
(R)-(+)-4-Benzyl-5,5-diphenyl-2-oxazolidinon
Formel: C₂₂H₁₉NO₂
Molecular Weight: 329,4 g/mol
CAS-Nummer: 191090-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR