3-Hydroxy-3-methylbuttersäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Hydroxyisovaleric acid, 3-Hydroxy-3-methylbutanoic acid

55453-1G
SIAL55453-1GEA 379 EUR
SIAL55453-1G
3-Hydroxy-3-methylbuttersäure, Sigma-Aldrich®
3-Hydroxy-3-methylbuttersäure
Formel: C₅H₁₀O₃
Molecular Weight: 118,13 g/mol
MDL: MFCD00059081
CAS-Nummer: 625-08-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR