[1,1'-Bis(diphenylphosphino)ferrocen]cobalt(II)chlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dichloro[1,1'-bis(diphenylphosphino)ferrocene]cobalt(II)

M-Clarity™ Qualitätsstufe = MQ100

555134-1G
SIAL555134-1GEA 70.8 EUR
SIAL555134-1G
[1,1'-Bis(diphenylphosphino)ferrocen]cobalt(II)chlorid, Sigma-Aldrich®
[1,1'-Bis(diphenylphosphino)ferrocen]cobalt(II)chlorid
Formel: C₃₄H₂₈Cl₂CoFeP₂
Molecular Weight: 684,23 g/mol
MDL: MFCD03427161
CAS-Nummer: 67292-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR