(4-Benzyloxyphenyl)essigsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-(4-(Benzyloxy)phenyl)acetic acid
M-Clarity™ Qualitätsstufe = MQ100
555398-1G
SIAL555398-1GEA
31
EUR
SIAL555398-1G
(4-Benzyloxyphenyl)essigsäure, Sigma-Aldrich®
(4-Benzyloxyphenyl)essigsäure
Formel:
(C₆H₅CH₂OC₆H₄)CH₂COOH Molecular Weight: 242.27 g/mol |
MDL:
MFCD00017540 CAS-Nummer: 6547-53-1 EINECS: 229-463-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...