4-(Trifluormethylthio)benzylamin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

555460-1G
SIAL555460-1GEA 104 EUR
SIAL555460-1G
4-(Trifluormethylthio)benzylamin, Sigma-Aldrich®
4-(Trifluormethylthio)benzylamin
Formel: C₈H₈F₃NS
Molecular Weight: 207,21 g/mol
MDL: MFCD00190129
CAS-Nummer: 128273-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR