3,5-Lutidin-N-oxid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,5-Lutidine N-Oxide, 3,5-Dimethylpyridine 1-oxide, 3,5-Dimethylpyridine N-Oxide

M-Clarity™ Qualitätsstufe = MQ100

555657-25G
SIAL555657-25GEA 293 EUR
SIAL555657-25G
3,5-Lutidin-N-oxid, Sigma-Aldrich®
3,5-Lutidin-N-oxid
Formel: C₇H₉NO
Molecular Weight: 123,1525 g/mol
MDL: MFCD00126839
CAS-Nummer: 3718-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR