2-Methoxy-4-nitrobenzolsulfonylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methoxy-4-nitrobenzolsulfochlorid, 2-Methoxy-4-nitrobenzolsulfonsäurechlorid, 2-Methoxy-4-nitrobenzol-1-sulfonylchlorid

M-Clarity™ Qualitätsstufe = MQ200

555932-5G
SIAL555932-5GEA 120 EUR
SIAL555932-5G
2-Methoxy-4-nitrobenzolsulfonylchlorid, Sigma-Aldrich®
2-Methoxy-4-nitrobenzolsulfonylchlorid
Formel: C₇H₆ClNO₅S
Molecular Weight: 251,65 g/mol
MDL: MFCD03094697
CAS-Nummer: 21320-91-2
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR