(4-Bromphenyl)methylsulfon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Brom-4-(methylsulfonyl)benzol

M-Clarity™ Qualitätsstufe = MQ100

556327-5G
SIAL556327-5GEA 56.3 EUR
SIAL556327-5G
(4-Bromphenyl)methylsulfon, Sigma-Aldrich®
(4-Bromphenyl)methylsulfon
Formel: C₇H₇BrO₂S
Molecular Weight: 235.1 g/mol
MDL: MFCD00025065
CAS-Nummer: 3466-32-8
EINECS: 222-421-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR