Dimethoxy(dimethyl)silan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Dimethyldimethoxysilan
M-Clarity™ Qualitätsstufe = MQ100
556688-25ML
SIAL556688-25MLEA
114
EUR
SIAL556688-25ML
Dimethoxy(dimethyl)silan, Sigma-Aldrich®
Dimethoxy(dimethyl)silan
Formel:
C₄H₁₂O₂Si Molecular Weight: 120,22 g/mol Siedepunkt: 82 °C (1013 hPa) Schmelzpunkt: –80 °C Dichte: 0,8638 g/cm³ (20 °C) |
MDL:
MFCD00025691 CAS-Nummer: 1112-39-6 EINECS: 214-189-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...