Bis(4-chlorphenyl)disulfid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bis(4-chlorophenyl) disulphide

M-Clarity™ Qualitätsstufe = MQ100

557161-25G
SIAL557161-25GEA 287 EUR
SIAL557161-25G
Bis(4-chlorphenyl)disulfid, Sigma-Aldrich®
Bis(4-chlorphenyl)disulfid
Formel: C₁₂H₈Cl₂S₂
Molecular Weight: 287,23 g/mol
MDL: MFCD00013642
CAS-Nummer: 1142-19-4
EINECS: 214-531-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR