5-Brom-2-thiophenboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Bromthiophen-2-boronsäure, 5-Brom-2-thienylboronsäure

M-Clarity™ Qualitätsstufe = MQ100

557684-5G
SIAL557684-5GEA 70.9 EUR
SIAL557684-5G
5-Brom-2-thiophenboronsäure, Sigma-Aldrich®
5-Bromo-2-thiopheneboronic acid
Formel: C₄H₄BBrO₂S
Molecular Weight: 206,85 g/mol
MDL: MFCD02094028
CAS-Nummer: 162607-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR