6-Chlorchinolin-2(1H)-on, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Chloro-2-hydroxyquinoline

M-Clarity™ Qualitätsstufe = MQ100

557773-5G
SIAL557773-5GEA 265 EUR
SIAL557773-5G
6-Chlorchinolin-2(1H)-on, Sigma-Aldrich®
6-Chlorchinolin-2(1H)-on
Formel: C₉H₆ClNO
Molecular Weight: 179,6030 g/mol
MDL: MFCD02930633
CAS-Nummer: 1810-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR