2-Chlorthioanisol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2-Chlorphenylmethylsulfid, 1-Chlor-2-(methylthio)benzol

M-Clarity™ Qualitätsstufe = MQ100

558001-5G
SIAL558001-5GEA 29.1 EUR
SIAL558001-5G
2-Chlorthioanisol, Sigma-Aldrich®
2-Chlorthioanisol
Formel: C₇H₇ClS
Molecular Weight: 158,65 g/mol
MDL: MFCD00040124
CAS-Nummer: 17733-22-1
UN: 3334
ADR: 9,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR