(R)-(-)-4-Penten-2-ol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-(-)-1-Penten-4-ol, (R)-(-)-Allyl-methylcarbinol

M-Clarity™ Qualitätsstufe = MQ100

558036-5G
SIAL558036-5GEA 717 EUR
SIAL558036-5G
(R)-(-)-4-Penten-2-ol, Sigma-Aldrich®
(R)-(-)-4-Penten-2-ol
Formel: C₅H₁₀O
MDL: MFCD03701536
CAS-Nummer: 64584-92-5
UN: 1987
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR