(S)-(+)-4-Phenyl-2-butanol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(S)-(+)-4-Phenylbutan-2-ol
M-Clarity™ Qualitätsstufe = MQ100
558095-1G
SIAL558095-1GEA
203
EUR
SIAL558095-1G
(S)-(+)-4-Phenyl-2-butanol, Sigma-Aldrich®
(S)-(+)-4-Phenyl-2-butanol
Formel:
C₁₀H₁₄O Molecular Weight: 150,22 g/mol |
MDL:
MFCD03093082 CAS-Nummer: 22148-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...