(R)-(-)-4-Phenyl-2-butanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-(-)-4-Phenylbutan-2-ol, (R)-4-Phenylbutan-2-ol

M-Clarity™ Qualitätsstufe = MQ100

558109-1G
SIAL558109-1GEA 159 EUR
SIAL558109-1G
(R)-(-)-4-Phenyl-2-butanol, Sigma-Aldrich®
(R)-(-)-4-Phenyl-2-butanol
Formel: C₁₀H₁₄O
Molecular Weight: 150,22 g/mol
MDL: MFCD03093081
CAS-Nummer: 39516-03-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR