Dipraseodymtrioxid, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

558249-10G
SIAL558249-10GEA 269 EUR
SIAL558249-10G
Dipraseodymtrioxid, Sigma-Aldrich®
Dipraseodymtrioxid
Formel: O₃Pr₂
Molecular Weight: 329,81 g/mol
MDL: MFCD00011178
CAS-Nummer: 12036-32-7
EINECS: 234-845-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR