4-Nitro-3-(trifluormethyl)benzolsulfonylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Nitro-3-(trifluoromethyl)benzene-1-sulphonyl chloride

M-Clarity™ Qualitätsstufe = MQ100

558575-5G
SIAL558575-5GEA 157 EUR
SIAL558575-5G
4-Nitro-3-(trifluormethyl)benzolsulfonylchlorid, Sigma-Aldrich®
4-Nitro-3-(trifluormethyl)benzolsulfonylchlorid
Formel: C₇H₃ClF₃NO₄S
Molecular Weight: 289,62 g/mol
MDL: MFCD03094387
CAS-Nummer: 39234-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR