1,4-Difluor-2-iodbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,5-Difluoroiodobenzene

M-Clarity™ Qualitätsstufe = MQ100

558591-5G
SIAL558591-5GEA 78.5 EUR
SIAL558591-5G
1,4-Difluor-2-iodbenzol, Sigma-Aldrich®
1,4-Difluoro-2-iodobenzene
Formel: C₆H₃F₂I
Molecular Weight: 239,99 g/mol
MDL: MFCD03094151
CAS-Nummer: 2265-92-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR