4-Brom-3-(trifluormethyl)benzolsulfonylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Brom-3-(trifluormethyl)benzol-1-sulfonylchlorid, 4-Brom-3-(trifluormethyl)benzolsulfochlorid, 4-Brom-3-(trifluormethyl)benzolsulfonsäurechlorid

M-Clarity™ Qualitätsstufe = MQ100

558621-1G
SIAL558621-1GEA 26 EUR
SIAL558621-1G
4-Brom-3-(trifluormethyl)benzolsulfonylchlorid, Sigma-Aldrich®
4-Brom-3-(trifluormethyl)benzolsulfonylchlorid
Formel: C₇H₃BrClF₃O₂S
Molecular Weight: 323,52 g/mol
Dichte: 1,833 g/cm³ (20 °C)
MDL: MFCD03094391
CAS-Nummer: 351003-47-9
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR