1-Chlor-2-fluor-3-iodbenzol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
558656-5G
SIAL558656-5GEA
30.7
EUR
SIAL558656-5G
1-Chlor-2-fluor-3-iodbenzol, Sigma-Aldrich®
1-Chloro-2-fluoro-3-iodobenzene
Formel:
C₆H₃ClFI Molecular Weight: 256,4439 g/mol |
MDL:
MFCD03094152 CAS-Nummer: 72373-82-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...