3-Chlor-2-fluorbenzolsulfonylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Chlor-2-fluorbenzol-1-sulfonylchlorid, 3-Chlor-2-fluorbenzolsulfochlorid, 3-Chlor-2-fluorbenzolsulfonsäurechlorid

M-Clarity™ Qualitätsstufe = MQ100

558672-1G
SIAL558672-1GEA 26 EUR
SIAL558672-1G
3-Chlor-2-fluorbenzolsulfonylchlorid, Sigma-Aldrich®
3-Chlor-2-fluorbenzolsulfonylchlorid
Formel: C₆H₃Cl₂FO₂S
Molecular Weight: 229,06 g/mol
MDL: MFCD03094234
CAS-Nummer: 351003-48-0
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR