5-Chlor-2-fluorbenzolsulfonylchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5-Chlor-2-fluorbenzolsulfonsäurechlorid, 5-Chlor-2-fluorbenzolsulfochlorid, 5-Chlor-2-fluorbenzol-1-sulfonylchlorid
M-Clarity™ Qualitätsstufe = MQ100
558680-1G
SIAL558680-1GEA
29.1
EUR
SIAL558680-1G
5-Chlor-2-fluorbenzolsulfonylchlorid, Sigma-Aldrich®
5-Chlor-2-fluorbenzolsulfonylchlorid
Formel:
C₆H₃Cl₂FO₂S Molecular Weight: 229,06 g/mol Schmelzpunkt: 41…44 °C |
MDL:
MFCD03094221 CAS-Nummer: 351003-49-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...